Zobell Solution, ORP Calibrator
Catalog # 66121-265
Supplier: YSI
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Bottle
- Description:Zobell Solution, ORP Calibrator
- Size:125 ml
- Environmentally Preferable:
- Cat. no.:66121-265
- Supplier No.:061320
Specifications
About this item
Zobell solution is used for calibrating ORP/Redox on several YSI instruments.
A convenient temperature chart with corresponding ORP values is included for simple calibration value inputs. Concentrated powder makes 125, 250, or 500 ml of calibration standard.