ContactUsLinkComponent
Budget Tabletop Training IV Stand
Budget Tabletop Training IV Stand
Catalog #: 470313-864
Simulator Training Iv Table Stand
Budget Tabletop Training IV Stand
Catalog #: 470313-864
Specifications
  • Description:
    Training Tabletop Stand
  • Cat. no.:
    470313-864
Specifications
About this item
A low cost stand for holding fluid supplies for simulators.
  • Holds blood or IV bags
  • Adjustable height
This two hook stand is a great accessory for any training program. Designed as a blood supply stand for a simulator, it can also be used to hold IV or blood bags for connection training. The stand has a 6" diameter base, and adjusts from 15" to 25" high. The base also has punch-outs, allowing users to attach suction cups (not included), or to attach the base to a surface using screws (not included) for additional stability when using at full extension.
Ordering information: Image may show items not included with purchase.
Customers Who Bought This Also Bought