Helix® 2065 Series Gloves, Hexarmor
Supplier: HexArmor
Certificates
About this item
Innovative Helix® series 2065 gloves feature a HPPE, fiberglass, and nylon blend shell with a seamless construction for enhanced comfort and breathability. A double-dipped flexible sandy nitrile palm coating provides 360° ANSI/ISEA level A3 cut protection and water resistance for wearers.
- Seamless construction of 13-gauge HPPE, fiberglass, and nylon blend shell for enhanced comfort and breathability
- Flexible sandy nitrile palm coating provides superior grip and abrasion resistance while also offering dexterity and tactility
- Knit wrist helps prevent dirt and debris from entering the glove
- Launderable for extended life
Helix® series gloves offer a wide variety of grip options to provide users with the appropriate grip for the task at hand. Providing coverage for any working condition, HexArmor is constantly researching and testing new formulas and application methods to maximize grip and function, helping workers redefine safety at their worksite.
Certifications: ANSI/ISEA Cut A3
Specifications
- Catalog No:
- 76318-194
- 76318-112
- 76318-116
- 76318-114
- 76318-118
- 76318-188
- 76318-192
- 76318-190
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Nylon/HPPE
- Nylon/HPPE
- Nylon/HPPE
- Nylon/HPPE
- Nylon/HPPE
- Nylon/HPPE
- Nylon/HPPE
- Nylon/HPPE
- Finish:
- Sandy Nitrile Palm
- Sandy Nitrile Palm
- Sandy Nitrile Palm
- Sandy Nitrile Palm
- Sandy Nitrile Palm
- Sandy Nitrile Palm
- Sandy Nitrile Palm
- Sandy Nitrile Palm
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist