Triethyl citrate 99.0-101.0% (alkalimetric, calculated on anhydrous substance) Ph. Eur., JPE, NF, SAFC®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Hydroxytricarballylic acid triethyl ester
Formula:C₁₂H₂₀O₇
MW:276.29 g/mol
Boiling Pt:294 °C (1013 hPa)
Melting Pt:–55 °C
Density:1.135 g/cm³ (25 °C)
MDL Number:MFCD00009201
CAS Number:77-93-0
:Storage Temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
Flash Pt:150 °C
:Boiling Pt:
:Boiling Pt: 294 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –55 °C
:Flash Pt:
:Flash Pt: 150 °C
:M.W. 276.29 g/mol
:Triethyl citrate
Storage Temperature:Ambient
EINECS:201-070-7
:294 °C (1013 hPa)
:150 °C
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (Alkalimetric, Calculated on Anhydrous Substance):99.0 - 101.0%
- Assay (GC, Calc. on Anhydrous Substance):99.0 - 100.5%
- Identity (IR-spectrum):Passes Test
- Identity (GC):Passes Test
- Identity (Wet Chemistry):Passes Test
- Identity (Refractive Index):Passes Test
- Appearance:Clear Solution, Not More Intense in Color than Matching Fluid A (1:10) and Reference Solution BY₆
- Acidity:Passes Test
- Refractive Index (n 20/D):1.440 - 1.446
- Refractive Index (n 25/D):1.439 - 1.441
- Density (d 25/25):1.135 - 1.139
- Density (d 20/20):1.135 - 1.145
- Viscosity (20 °C):26 - 38 mPa∙s
- Sulfate (SO₄):≤150 ppm
- Heavy metals (as Pb):≤5 ppm
- As (Arsenic):≤2 ppm
- Cyclohexane (HS-GC):≤3880 ppm
- Ethanol (HS-GC):≤5000 ppm
- Other Residual Solvents (ICH (Q3C)):Excluded by Manufacturing Process
- Related Substances (GC) (Largest Single Impurity):≤0.2%
- Related Substances (GC) (Sum of All Impurities):≤ 0.5 %
- Triethyl Aconitate (GC):≤0.2%
- Related substances (GC):Passes Test
- Sulfated ash (600 °C):≤0.1%
- Water (According to Karl Fischer):≤0.25%
Specification Test Results
Customers Who Bought This Also Bought