Manual/Press, Spex SamplePrep
Catalog #: 76181-928
Supplier: SPEX
Specifications
- Description:Press
- W×D×H:91×41×38 cm (36×16×15")
- Weight:68 kg (150 lbs.)
- Cat. no.:76181-928
- Electrical:115 V, 60 Hz
Specifications
About this item
Full-size 12-ton (10.9 metric ton) Carver® hydraulic laboratory pellet press that accepts 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets.
- Accepts all SPEX SamplePrep evacuable pellet die sets
- Can be operated anywhere in the world, even under the most primitive conditions
- Economical alternative to motorized presses for labs with a small sample load
- Hardened steel buffer plate is engraved with concentric circles and enables easy centering of the die
- Manual, hydraulic laboratory press with buffer plate
- Rugged, reliable and noted for its extraordinary durability
- Upper platen is equipped with a safety shield for operator protection
Manually controlled. Ideal for pressing sample pellets for XRF, IR, and other analytical techniques. Typical samples include: cement, rocks, minerals, soils, ceramics, pharmaceuticals.
Certifications: CE Approved.
Customers Who Bought This Also Bought