pH Buffer Solutions, Referenced at 20 °C, Traceable to NIST, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
Certificates
About this item
- Citrate and phosphate buffer solutions contain fungicide
- Traceable to SRM from NIST
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.