ContactUsLinkComponent
Quanta-Ray Dye Laser Flow Through Cells, FireFlySci
Quanta-Ray Dye Laser Flow Through Cells, FireFlySci
Catalog #: 76015-076
Supplier:  FIREFLYSCI, INC.
Quanta Ray Dye Laser Flow Through Cell Type 0005-0023-2 6x12mm
Quanta-Ray Dye Laser Flow Through Cells, FireFlySci
Catalog #: 76015-076
Supplier:  FIREFLYSCI, INC.
Specifications
  • Description:
    Dye Laser Flow Through Cell
  • Material:
    UV Quartz
  • Lightpath:
    6×12 mm
  • Cat. no.:
    76015-076
Specifications
About this item
Dye laser cells are manufactured with extreme surface flatness extending to the edges of the cell.
  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Each cell goes through a rigorous inspection process before leaving our factory
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in UV Quartz
  • Available in dual 6×12 mm lightpath
  • Four windows optically polished
  • Open top and open bottom flow through cell
Flatness is checked via interferometer. Premium quality Firefly cuvettes are made with the best quality quartz and glass material available transmission values.
Customers Who Bought This Also Bought