About this item
High-quality premium vinyl upholstered stack chairs available in choice of institutional grade fabrics or thick 0.80 mm vinyl.
- Square-tube 7/8" gauge steel frame 18 with 5/8" under seat and H-braces
- Steel standard: ASTM A513
- Foam standard: 1.8 lbs. density, 46 ILD
- Thick grade a foam 2"; cushion is double stitched with a spacious waterfall seat
- MDF core seat and back
- 10 year warranty
The 9200 Dome-Back stack chair is durable and classy enough to use in the busiest social setting, be it a dining room or corporate banquet hall. It features a strong square 18 gauge steel frame plus an under-seat H-shaped brace for extra support. The seat features supremely comfortable and spacious 2" thick high-density foam padding with easy-to-clean vinyl upholstery or institutional grade fabric, and a waterfall-style edge. Plywood core within both the seat and rounded backrest which has a built-in handle for easy moving and stacking. Fortifying stacking bars and twelve plastic bumpers protect the powder-coated paint finish from scratches when the chair is stacked (up to ten units high). Patterned fabrics rated to 67000 double rubs, fabric and foam are Cal-117 rated. The back has a convenient handhold for easier moving and stacking. Stacking bars and 12 plastic stack bumpers ensure stacking securely without damaging the attractive powder-coated frame finish. Stack 8 to 10 high with DY81 or DY9000 dollies. Solid fabrics rated to 130000 double rubs.
Specifications
- Catalog No:
- 76276-436
- 76276-434
- 76276-456
- 76276-454
- 76276-446
- 76276-444
- 76276-442
- 76276-736
- 76276-448
- Model:
- 76276-436
- 76276-434
- 76276-456
- 76276-454
- 76276-446
- 76276-444
- 76276-442
- 76276-736
- 76276-448
- Frame colour/finish:
- Black Sandtex
- Silvervein
- Sandtex
- Silvervein
- Silvervein
- Black Sandtex
- Silvervein
- French Beige
- Black Sandtex
- Seat/back color:
- Midnight Blue
- Midnight Blue
- Ebony Black
- Ebony Black
- Midnight Blue
- Panther Black
- Panther Black
- French Beige
- Midnight Blue