Coomassie® brilliant blue R-250 solution electrophoresis gel stain, ready-to-use
Catalog #: CA95043-426
Supplier: G-Biosciences
CAS Number: 6104-59-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 L
- Cat. no.:CA95043-426
- Supplier No.:786-498
Specifications
Specification Test Results
- Coomassie® brilliant blue R-250 solution electrophoresis gel stain, ready-to-use:
Specification Test Results
About this item
Synonyms: Serva Blue R,Brillant blue R,Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium salt,Coomassie Blue R 250,Brilliant Cyanine 6B,Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt,Acid Blue 83,Brilliant Blue R
:Coomassie® brilliant blue R-250
:Storage Temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 174 to 180 °C
:Flash Pt:
:Flash Pt:
:M.W. 825.98 g/mol
Formula:C₄₅H₄₄N₃NaO₇S₂
MW:825.98 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00041762
CAS Number:6104-59-2
EINECS:228-058-4
Customers Who Bought This Also Bought