TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Tris(hydroxymethyl)methylamine hydrochloride,2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride,Trizma® hydrochloride,Trizma(R) hydrochloride,Tris(hydroxymethyl)aminomethane hydrochloride,TRIS HCl,Tromethane hydrochloride,Tris(hydroxymethyl)methylammonium chloride,Trometamol hydrochloride
Formula:NH₂C(CH₂OH)₃·HCl
MW:157.6 g/mol
MDL Number:MFCD00012590
CAS Number:1185-53-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 150 to 152 °C
:Flash Pt:
:Flash Pt:
:M.W. 157.6 g/mol
:TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
:GHS07
:M.O.C. Category R
:12,09902
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Assay (NH₂C(CH₂OH)₃·HCl):≥99.0 %
- Appearance (colorless crystals to white crystalline powder):Passes Test
- Absorbance of a 1M Solution - 260 nm:≤ 0.06
- Absorbance of a 1M Solution - 280 nm:≤ 0.06
- Absorbance of a 1M Solution - 400 nm:≤ 0.01
- Identification (by IR):Passes Test
- Loss on Drying at 105 °C:≤ 0.5 %
- Water (H₂O)(by Karl Fischer titrn):≤ 0.5 %
- Endotoxin Concentration (EU/g):≤ 2.5
- Melting Point:147 - 153 °C
- pH of 0.5M Solution at 25 °C:3.5 - 5.0
- Enzyme Activity - DNase Activity:Passes Test
- Enzyme Activity - RNase Activity:Passes Test
- Enzyme Activity - Protease Activity:Passes Test
- Trace Impurities - Sodium (Na):≤ 50.000 ppm
- Trace Impurities - Aluminum (Al):≤ 5.000 ppm
- Trace Impurities - Barium (Ba):≤ 5 ppm
- Trace Impurities - Bismuth (Bi):≤ 5 ppm
- Trace Impurities - Cadmium (Cd):≤ 5.000 ppm
- Trace Impurities - Calcium (Ca):≤ 10.0 ppm
- Trace Impurities - Cobalt (Co):≤ 5.000 ppm
- Trace Impurities - Chromium (CR):≤ 5 ppm
- Trace Impurities - Potassium (K):≤ 50 ppm
- Trace Impurities - Lithium (Li):≤ 5 ppm
- Trace Impurities - Manganese (Mn):≤ 5 ppm
- Trace Impurities - Molybdenum (Mo):≤ 5 ppm
- Trace Impurities - Nickel (Ni):≤ 5.000 ppm
- Trace Impurities - Arsenic (As):≤ 1 ppm
- Trace Impurities - Copper (Cu):≤ 5 ppm
- Trace Impurities - Iron (Fe):≤ 5 ppm
- Trace Impurities - Lead (Pb):≤ 1 ppm
- Trace Impurities - Magnesium (Mg):≤ 5 ppm
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Filtered through a 0.2 micron filter.:
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR4107
SDS
spec_JT4107-07 spec_JT4107-09 spec_JT4107-F7 spec_JT4107-F9
Product specification
Customers Who Bought This Also Bought