J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Phosphate
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
This BAKER ANALYZED® Reagent is a phosphate buffer solution for laboratory, research, and/or manufacturing uses.
- Contains no antimicrobial preservative
Each lot is compared to NIST Standard Reference Buffer.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.