Sodium hydroxide 97.5-100.5% (by acidimetry), pellets, J.T.Baker®
Catalog #: CAJT5565-28
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 1310-73-2
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:25 kg
- Cat. no.:CAJT5565-28
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (NaOH) (by acidimetry):97.5 - 100.5 %
- ACS - Sodium Carbonate (Na₂CO₃):≤1.0 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Chloride (Cl):≤0.005 %
- ACS - Heavy Metals (as Ag):≤0.002 %
- ACS - Potassium (K):≤0.02 %
- ACS - Nitrogen Compounds (as N):≤0.001 %
- ACS - Phosphate (PO₄):≤0.001 %
- ACS - Sulfate (SO₄):≤0.003 %
- ACS - Iron (Fe):≤0.001 %
- ACS - Magnesium (Mg):≤0.002 %
- ACS - Mercury (Hg):≤0.100 ppm
- ACS - Nickel (Ni):≤0.001 %
- EP/BP - Characteristics:Passes Test
- EP/BP - Identification:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Heavy Metals (as Pb):≤20 ppm
- EP/BP - Iron (Fe):≤10 ppm
Specification Test Results
About this item
Synonyms: Caustic soda
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97,5-100,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 323 °C
:Flash Pt:
:Flash Pt:
:M.W. 40 g/mol
:323
:1390
UN:1823
:8
:II
:GHS05
:12,08772
:14,08627
Formula:NaOH
MW:40 g/mol
MDL Number:MFCD00003548
CAS Number:1310-73-2
Customers Who Bought This Also Bought