Sodium phosphate dibasic, anhydrous USP, Multi-Compendial HP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: sec-Sodium phosphate,di-Sodium hydrogen orthophosphate,di-Sodium hydrogen phosphate anhydrous,Sodium hydrogen phosphate,Disodium phosphate
Formula:Na₂HPO₄
MW:141.96 g/mol
Melting Pt:250 °C
Density:1.53 g/cm³ (20 °C)
MDL Number:MFCD00003496
CAS Number:7558-79-4
:Storage Temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 250 °C
:Flash Pt:
:Flash Pt:
:M.W. 141.96 g/mol
:Sodium phosphate dibasic
Storage Temperature:Ambient
EINECS:231-448-7
REACH:01-2119489797-11
:M.O.C. Category R
:12,08805
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- Meets JPE Specifications:
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- Meets E.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (Na₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Identification:Passes Test
- USP - Loss on Drying at 130°C:≤ 5.0 %
- USP - Insoluble Substances:≤ 0.4 %
- USP - Chloride (Cl):≤ 0.06 %
- USP - Sulfate (SO₄):≤ 0.2 %
- USP - Arsenic (As):≤ 16 ppm
- JPE - Assay (dried basis):≥ 99.0 %
- JPE - Arsenic (As):≤ 2 ppm
- JPE - Carbonate:Passes Test
- JPE - Chloride (Cl):≤ 0.011 %
- JPE - Clarity and Color of Solution:Passes Test
- JPE - Heavy Metals:≤ 10 ppm
- JPE - pH (1-100):9.0 - 9.4
- JPE - Sulfate (SO₄):≤ 0.038 %
- JPE - Loss on Drying:≤ 2.0 %
- EP - Assay (Na₂HPO₄) (dried basis):98.0 - 101.0 %
- EP - Appearance of Solution:Passes Test
- EP - Identification A:Passes Test
- EP - Identification B:Passes Test
- EP - Identification C:Passes Test
- EP - Identification D:Passes Test
- EP - Reducing Substances:Passes Test
- EP - Chloride (Cl):≤ 200 ppm
- EP - Iron (Fe):≤ 20 ppm
- EP - Loss on Drying:≤ 1.0 %
- EP - Monosodium Phosphate:≤ 2.5 %
- EP - Sulfate (SO₄):≤ 500 ppm
- Endotoxin Concentration (EU/g):≤ 2.5
- Microbial Testing – Total Aerobic Microbial Count (USP):≤ 10 cfu/g
- Microbial Testing – Total Yeast and Mold Count (USP):≤ 10 cfu/g
- Microbial Testing - Salmonella (USP):Negative
- Microbial Testing - Escherichia coli (USP):Negative
- Microbial Testing - Staphylococcus aureus (USP):Negative
- Microbial Testing - Pseudomonas aeruginosa (USP):Negative
- Microbial Testing -Bile tolerant gram-negative bacteria:Negative
- Microbial Testing – Clostridia:Negative
- Microbial Testing - Candida albicans:Negative
- Trace Impurities - Cadmium (Cd):≤ 200 ppb
- Trace Impurities - Lead (Pb):≤ 500 ppb
- Trace Impurities - Arsenic (As):≤ 1500 ppb
- Trace Impurities - Mercury (Hg):≤ 300 ppb
- Trace Impurities - Cobalt (Co):≤ 500 ppb
- Trace Impurities - Vanadium (V):≤ 1000 ppb
- Trace Impurities - Nickel (Ni):≤ 2000 ppb
- Trace Impurities - Thallium (Tl):≤ 800 ppb
- Trace Impurities - Gold (Au):≤ 10000 ppb
- Trace Impurities - Palladium (Pd):≤ 1000 ppb
- Trace Impurities - Iridium (Ir):≤ 1000 ppb
- Trace Impurities - Osmium (Os):≤ 1000 ppb
- Trace Impurities - Rhodium (Rh):≤ 1000 ppb
- Trace Impurities - Ruthenium (Ru):≤ 1000 ppb
- Trace Impurities - Selenium (Se):≤ 8000 ppb
- Trace Impurities - Silver (Ag):≤ 1000 ppb
- Trace Impurities - Platinum (Pt):≤ 1000 ppb
- Trace Impurities - Lithium (Li):≤ 25000 ppb
- Trace Impurities - Antimony (Sb):≤ 9000 ppb
- Trace Impurities - Barium (Ba):≤ 70000 ppb
- Trace Impurities - Molybdenum (Mo):≤ 150000 ppb
- Trace Impurities - Copper (Cu):≤ 30000 ppb
- Trace Impurities - Tin (Sn):≤ 60000 ppb
- Trace Impurities - Chromium (Cr):≤ 110000 ppb
- No Class 1,2,3 or other solvents are used or produced in the:
- J.T.Baker®, Sodium phosphate dibasic, anhydrous USP, Multi-Compendial HP:
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR4953
SDS
spec_JT4953-01 spec_JT4953-07 spec_JT4953-65
Product specification
Customers Who Bought This Also Bought