KoolTemp® GTS-52 Urethane Insulated Shipper, Cold Chain Technologies
Catalog #: 89177-022
Supplier: Cold Chain Technologies
Small Business Enterprise
Specifications
- Description:GTS-52 urethane insulated shipper
- Payload Dimensions:3¾L×3¾W×8⅝H"
- Cat. no.:89177-022
- Exterior Dimensions:197/8L×197/8W×21"H
- Supplier No.:GTS-52-SU
Specifications
About this item
The KoolTemp® GTS-52 urethane insulated shipper maintains small payloads from 2 to 8 °C for 96 hours under extreme weather conditions. The simple universal pack out minimizes costs, including labor and turnaround time.
- Non-ozone depleting
- Lightweight, easy universal packout
The GTS-52 urethane insulated shipper is an excellent thermal solution for long-term domestic and international shipments of valuable vaccines, pharmaceutical products, and blood. The entire container is thermally sealed when the hard cover is fit securely onto the base. The modular design allows for reuse under proper conditions, and replacement when necessary.
Customers Who Bought This Also Bought