To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Plant samples
- Target molecule:RNA
- Description:Quick-RNA plant kit
- Size:50 Preps
- Description:Quick-RNA plant kit
- Yield:50 µg RNA (binding capacity), ≥25 µl (elution volume)
- Cat. no.:76020-448
Specifications
About this item
Quick-RNA plant miniprep kit is designed for rapid isolation of RNA from various plant samples such as leaves, stems, buds, flowers, fruits and seeds.
- Extract intact high-quality total RNA (including small/micro RNAs) from a wide variety of plant samples
- Ultra-high density bashing bead technology for complete disruption and sample lysis
- Inhibitor free RNA is ready for all downstream applications
- Size Range Total RNA ≥ 17 nt
The Zymo-Spin IIICG column allows for high-capacity DNA elimination and the subsequent Zymo-Spin IIC column efficiently adsorbs total RNA. The RNA is washed and then eluted with DNase/RNase free water. For inhibitor removal, the eluted RNA can be treated by running the sample through the Zymo-Spin III-HRC spin filters. The RNA is suitable for use in various subsequent procedures including sequencing, RT-PCR and microarray.