Microinjection Fish Molds, IBI Scientific
Catalog #: 71002-572
Supplier: IBI Scientific
Specifications
- Description:Microinjection fish molds
- Cat. no.:71002-572
- Supplier No.:FM-600
Specifications
About this item
These microinjection zebra fish mold has six angled lanes to hold zebra fish embryos in place during microinjection.
- Forms six angled lanes in an agarose slurry
Mold has a plastic handle on top.
Customers Who Bought This Also Bought