About this item
Thermal-transfer printable tags for the identification of high-profile PCR tubes, strips and plates, it allows PCR tubes to be labeled with far more information, in a fraction of the time.
- Versatile design allows PCR tubes, strips, and plates to be tagged in a variety of configurations and styles
- Quick, bird’s eye view of all sample information
- Identify PCR samples with more information than cap dots and tube side rectangular labels combined
- Thermal-transfer printable, allows printing with barcodes for easy tracking
- They are safe for use in thermocyclers, and long-term storage in laboratory refrigerators and freezers
PCR-TagTrax™ offers an innovative and helpful approach to identifying, tracking, and storing 0.2 ml high-profile PCR tubes, high-profile eight tube PCR strips, as well as 96-well high-profile PCR plates (non-skirted). With an adhesive-free, glove-friendly design, tags are able to withstand a wide range of temperatures (–196 to 150 °C).
Thermal-transfer printable, the tags provide a means to identify PCR tubes and strips with alphanumeric text, as well as 1D and/or 2D barcodes. For use in biomedical research labs, genetic analysis facilities (genotyping, sequencing, cloning), genealogy and family records service companies, microbial sequencing service companies, as well as forensic laboratories.