Specimen Containers with Snap Cap, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
Disposable containers for specimen collection.
- Cap and container are made of polypropylene
- Disposable
- Tight fitting lids
These disposable containers made of polypropylene are ideal for collection and storage of fluids, powders, solids, pathology specimens, and hazardous samples.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.