PowerCoat® PVC Chemical Resistant Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Protects hands in hazardous liquid applications. Triple layer PVC coating is resistant to oils, chemicals, and many acids and solvents.
- Coating is soft and flexible
- Comfort and flexibility reduces hand fatigue
- Rough grip
- Seamless cotton knit liner
Certifications: EN 388: 4121 mechanical hazards. EN 374-3: JKL chemical hazards.
Specifications
- Catalog No:
- CA46100-496
- CA46100-498
- CA46100-500
- CA46100-502
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Material:
- PVC
- PVC
- PVC
- PVC