Extended Cuff Disposable Nitrile Gloves, Microflex®, Ansell
Supplier: Microflex
Certificates
About this item
Extra long cuff for superior forearm protection, and extra security against cross contamination.
- Textured fingers and palm ensure a secure grip in a variety of applications
- Offers greater puncture resistance and chemical resistance than natural rubber latex
Made with soft nitrile formulation for comfortable fit and feel while improved cuff design helps prevent gloves from rolling down.
Specifications
- Catalog No:
- CA75795-898
- CA75795-894
- Model:
- 93-243
- 93-243
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Cuff style:
- Beaded
- Beaded