Synthetic Vinyl Medical Examination Gloves, Powder Free, Bio Nuclear Diagnostics
Supplier: Bio Nuclear Diagnostics
Certificates
About this item
Single use medical examination glove, made of stretchy synthetic vinyl.
- Latex-free with the elasticity of latex
- High dexterity and tactile sensitivity
- Beaded cuffs to prevent rolling back and tearing
- Health Canada Approved
Suitable for general patient care, household activities, handling food or other non-corrosive materials.
Certifications: Meets all ASTM standards.
Specifications
- Catalog No:
- CA32902-034
- CA32902-032
- CA32902-042
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Food contact approved:
- Yes
- Yes
- Yes