541 Flat Dipped Cut Resistant Glove, Showa
Supplier: Showa-Best Glove
Certificates
About this item
Using revolutionary flat-dipped technology, this glove empowers the user with high grip and is designed to hide grime.
- Cut-resistant
- HPPE liner with seamless 13 gauge
- Flat-dipped, gray colored PU coating
- Rough grip
- Elastic knit-wrist
- Launderable
It provides high degree of comfort when performing delicate tasks in work environments that are greasy and risk of cuts is high.
Certifications: CE Cat. II marked. EN 388:2003-4342. ANSI / ISEA 105-2016 CUT A2. ANSI / ISEA 105-2016 PUNCTURE 3.
Specifications
- Catalog No:
- CA94024-054
- CA94024-052
- CA94024-050
- CA94024-048
- CA94024-046
- Model:
- 541
- 541
- 541
- 541
- 541
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Cuff style:
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff