ZORB-IT® Oily Grip Cut Resistant Gloves, Showa
Supplier: Showa
Certificates
About this item
These cut resistant gloves provides extra grip in oily conditions due to an extra absorbent nitrile coating.
- Cut resistance level D
- Kevlar® liner for improved cut resistance
- Kevlar® liner is inherently flame resistant
- Seamless knitted liner prevents irritation
- Lightweight
Manufactured using patented hand former design for a comfortable fit and to minimize hand fatigue.
The 15 gauge breathable liner allows warm air and moisture from inside to escape, keeping hands dry and comfortable.
Sponge nitrile palm absorbs oil and grease to increase grip and durability in oily environments.
Certifications: CE Cat. II marked. EN 388: 2016-4X32D.
Specifications
- Catalog No:
- 76264-486
- 76264-484
- 76264-482
- 76264-504
- 76264-502
- Model:
- 4561
- 4561
- 4561
- 4561
- 4561
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Liner material:
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®
- Material:
- Nitrile/Kevlar®
- Nitrile/Kevlar®
- Nitrile/Kevlar®
- Nitrile/Kevlar®
- Nitrile/Kevlar®
- Finish:
- Sponge Nitrile Palm
- Sponge Nitrile Palm
- Sponge Nitrile Palm
- Sponge Nitrile Palm
- Sponge Nitrile Palm
- Gauge:
- 15
- 15
- 15
- 15
- 15