Food Safe Nitrile Gloves, Showa
Supplier: Showa
Certificates
About this item
These gloves are liquid-proof and chemical resistant and are ideal for professionals working in food industries.
- Lightweight
- Comfortable second skin feel
- Unflocked to prevent the risk of food contamination
- Fishscale pattern on palm and back of hands
- Extremely strong
- Quick and easy donning
Suitable for use with all food categories for short term, repeated contact.
Blue color allows for immediate identification of broken pieces, eliminating risk of contamination.
Certifications: Cat. III marked. EN 374-1: 2016 / Type B. EN 374-5: 2016-VIRUS. Directive 2002/72 EC.
Customers Who Bought This Also Bought