TN300B Series Class 100 Nitrile Glove, TechNiGlove
Supplier: Techniglove
Certificates
About this item
TechNiCleaned Class 100 nitrile gloves are puncture-resistant and 100% nitrile, reducing the potential for allergic reactions to natural ruber protiens.
- Blue finish
- Batch specific COC available
These products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary microtextured formers offer an excellent grip on all surfaces. These gloves are suitable for use in cleanrooms and contamination controlled work environments, pharmaceutical and medical device manufacturing.
Packaging: Packaged 100 gloves per double poly-sealed bag, 10 bags per case.
Specifications
- Catalog No:
- 76302-858
- 76302-860
- 76302-852
- 76302-850
- 76302-856
- 76302-854
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Microtextured Fingertips
- Cuff length:
- 9.5"
- 9.5"
- 9.5"
- 9.5"
- 9.5"
- 9.5"
Specifications
Related Information
TechNiCleaned Pharmaceutical Class 100 Nitrile Gloves TechNiGlove
Product Literature
Customers Who Bought This Also Bought