Arctic Winter Gloves, Portwest
Supplier: Portwest
Certificates
About this item
Arctic Winter Gloves are specially designed for use in cold conditions. Twin liner traps heat through increased insulation.
- ³/₄ dipped for increased protection
- 100% breathable seamlessnylon lining
Flexible sandy nitrile coating offers great grip in wet and dry conditions.
Certifications: AS/NZS 2161 .5. AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 CUT Level (A2). EN 420. EN 388:2003 - 4.1.4.1. EN 511 0.2.X. cold protection.
Specifications
- Catalog No:
- 76103-488
- 76103-490
- 76103-480
- 76103-482
- 76103-484
- 76103-486
- Model:
- A146
- A146
- A146
- A146
- A146
- A146
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Sandy 3/4 Coating
- Sandy 3/4 Coating
- Sandy 3/4 Coating
- Sandy 3/4 Coating
- Sandy 3/4 Coating
- Sandy 3/4 Coating