Eco Best® Biodegradable, Chemical Resistant, Hi-Vis Gloves, Showa
Supplier: Showa
Certificates
About this item
Complete biodegradable nitrile disposable glove with Eco Best Technology® in a hi-vis orange color protects against oils, hydrocarbons, grease and many chemicals.
- Fits like second skin
- Fluorescent orange color increases visibility
- Bisque finish on palm and fingers provides a long-lasting grip
- Impermeable, for working in damp or greasy environments
- Rolled cuff prevents dirt from entering the glove
- Chlorinated for ease of donning and doffing
Certifications: CE Cat. III marked. EN388:2016, 1001X. EN374-1:2016/Type A. EN374-5:2016. Directive 2002/72 EC.
Specifications
- Catalog No:
- 76264-494
- 76264-492
- 76264-496
- 76264-490
- 76264-488
- 76264-520
- Model:
- 707HVO
- 707HVO
- 707HVO
- 707HVO
- 707HVO
- 707HVO
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Cuff style:
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Rolled
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes