Specifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove size:L
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Cuff style:Gauntlet
- Cuff length:4.5"
- Cat. no.:CA76034-610
Specifications
About this item
This style of Endura® glove is made from smooth cowgrain leather for dexterity and some oil and water resistance. This is a heavier leather compared to other types, providing increased durability and abrasion resistance.
- Grain cowhide offers superb abrasion protection
- Gunn-cut styling
- Double Kevlar® stitching for added durability
- Strapped thumb for rugged durability
- Split-leather cowhide gauntlet cuffs for extra wrist protection
- Out-seam sewn for reduced rubbing and chafing
All-leather construction with strapped thumbs and double-sewn forefinger. Complete with 4.5" split-leather cuffs and seams that are positioned toward the back of fingers for reduced rubbing and annoyance on top of longer seam life.
Glove color: Cream