Synthware Filter Funnel, Kemtech America
Catalog #: 60004-204
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Porosity:Medium
- ST Joint Size:14/20
- Disc O.D.:20 mm
- Supplier No.:F381420M
Specifications
About this item
This filter funnel is made of glass, and with a fritted disc.
- Microscale
- heavy wall borosilicate construction
- Medium porosity fritted disc
- Drip tip
This fritted filter funnel is perfect for microscale filtration applications.
Customers Who Bought This Also Bought