Large HDPE Funnels, BrandTech® Scientific
Supplier: Brandtech
Certificates
About this item
Large funnels, available in two sizes and stem diameters to fit most applications.
- High density polyethylene
- Large sizes
- 89400-738 features handle
These VITLAB® large funnels are made of heavy-duty high-density polyethylene for durability and chemical resistance. Upper section has a large diameter, and large volume capacity to help prevent spillage. 60˚ angle allows rapid flow.
Customers Who Bought This Also Bought