Specifications
- Description:Sewable LED Moons
- Cat. no.:470311-288
Specifications
About this item
Color changeable sewable moons. Add cool decorations and color to your interactive textile projects. These moons can be used without a microcontroller but requires a sewable battery holder. Use it with your LilyPad or other textile ready microcontroller.
- RGB programmable colors
- Fun design to go along with the sewable stars
- Resistors are on the board
- Larger size so they can be easily sewn
These cool LEDs have a built-in resistor. They rely on conductive thread. Our high-quality conductive thread is a perfect companion to these sewable moons. With the battery removed, fabrics with electronics can be washed. Conceptually, these circuits are identical to their wire-and-solder counterparts, observing the same principles of polarity and resistance. Only the materials, methods and sometimes scale are different. There is a lot more to sewable circuits than LEDs. Check out the Sparkle Labs E-Textile kit.