Corning® Polycarbonate Erlenmeyer Flask, Male Luer Lock, Corning
Supplier: Corning
Certificates
About this item
Corning closed system Erlenmeyer flasks will both reduce contamination risk and enhance the productivity of your shaker culture operations when adding or removing medium and cells.
- Unit is sterilized by gamma radiation
- Nonpyrogenic and RNase-/DNase-free
- Molded-in graduations for accuracy (±5%)
- Male luer lock disconnect
- Sterility assurance level (SAL) of 10–6
Flask is made from optically clear polycarbonate (USP Class VI material) and is preassembled with an aseptic transfer cap. It also has chemically resistant, heat-sealable, heat-sealable, flexible tubing, ( I.D×O.D: ¹/₈×¹/₄") of length 36" with connector.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
- Catalog No:
- 89235-032
- 30621-424
- 89511-300
- 89511-304
- 89511-302
- 89511-306
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Specifications
Related Information
Certificates
Search by Lot Number30621-424_35319035 89511-300_34719046 89511-304_35119010
Conformity declaration
Customers Who Bought This Also Bought