Splicing and Maintenance Chaps, Series 9911, HexArmor
Supplier: HexArmor
Certificates
About this item
These HexArmor Series 9911 Chaps protect the body from high-risk hazards on the job site. In addition to high puncture-resistance, these full-length chaps provide ISEA Level 5 cut resistance.
- Cut, puncture and abrasion resistant, with front-to-back leg coverage
- Three layers of protection on knee (front only)
- Reflective tape for visual awareness
- Launderable
Made with SuperFabric® brand material for some of the highest cut-resistance available in the industry (ANSI/ISEA level A7 and CE level 5).
Certifications: ANSI/ISEA A7, EN 388 4532.
Specifications
- Catalog No:
- 76208-238
- 76208-112
- 76208-134
- 76208-136
- 76208-130
- 76208-132
- Model:
- 9911
- 9911
- 9911
- 9911
- 9911
- 9911
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis