TLG® Sure-Grip Separatory Funnels, Sati International
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
Globe-shaped separatory funnel with a long stopper neck and straight section between the funnel body and stopcock, which provides a secure grip while shaking and also aids in clamping. Supplied complete with the listed PP or hollow glass stopper.
- Available with Teflon® stopcock
- Stopcock bore: 4 mm
- Stopper No. 27
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Annealing point: 560 °C
Linear coefficient of expansion: 32×10-7/°C
Softening point: 820 °C
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”
Customers Who Bought This Also Bought