TLG® Jacketed Buchner Filter Funnels, Sati International
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
Jacket encloses entire funnel bowl and upper part of stem. Circulation of liquid through the jacket permits warming or cooling of funnel and contents during filtration.
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Annealing point: 560 °C
Linear coefficient of expansion: 32×10-7/°C
Softening point: 820 °C
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”
Customers Who Bought This Also Bought