TLG® Funnels, Solvent Addition, Sati International
Supplier: CHEM SCIENCE INC CA
Certificates
About this item
Design of funnel permits addition of solvents to flash chromatography columns without disturbing the silica gel/packing layer.
- Lower end of funnel is closed
- Series of holes throughout stem to drain solvent
- Top of funnel is 90 mm diameter
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Annealing point: 560 °C
Linear coefficient of expansion: 32×10-7/°C
Softening point: 820 °C
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”
Customers Who Bought This Also Bought