Gas Cylinder Barricade Rack, Justrite®
Supplier: Justrite
Certificates
About this item
These Gas Cylinder Barricade Racks provide safety when moving and storing of gas cylinders.
- Secure cylinders up to 305 mm (8 - 12") in diameter
- For indoor or outdoor bulk cylinder storage
- Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
- Barricade racks feature two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
Recommended for indoor or outdoor bulk cylinder storage, barricade racks surround cylinders with 51 mm (2") square steel structural tubing, continuously seam welded at all junction points and sealed with exterior grade powder-coat paint for long lasting strength, durability, and dependability.
Pre-drilled 14 mm (0.531") diameter floor mounting holes included on lower tube structure.
Certifications: Barricade racks are NFPA 1 and NFPA 55 approved, OSHA approved, CGA-P1 approved, and High Hazard Seismic Zone approved.
Meets USGS high-hazard seismic zone requirements when properly mounted.
Meets USGS high-hazard seismic zone requirements when properly mounted.
Ordering information: For seismic installations, add Model 35350 S4-rated anchor bolts (not included).
Frequently Bought Together