RS-P28 Printer, METTLER TOLEDO®
Catalog #: 97016-772
Supplier: Mettler Toledo
Specifications
- Description:RS-P28 Printer
- Cat. no.:97016-772
- Print Speed:2.3 lines/s
- Supplier No.:11124324
- Paper Type:Standard paper
Specifications
About this item
The printer supports the connected instrument languages, whether it operates in English, Russian, Chinese (balance), Japanese or Latin charaters.
- Statistical and totaling applications
- Fulfills GxP requirements
- RS232 interfaces
- Dot matrix printing technology
Build in real time and date allows users to record all measurements with customized data printouts.
Customers Who Bought This Also Bought