DURAN® HPLC Bottles, GL 45, DWK LIFESCIENCES
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Complete system comprising of DURAN® pressure plus laboratory bottle with four port screw cap (PP), four screw connections (black, M8 thread) and silicone seals.
- Easy-to-read scale and large labelling field for easy marking
- Unused ports can be sealed with silicone blanking seals
- Autoclavable at 121 °C
Connection of different hose diameters (1.6 and 3.2 mm) as well as sterile pressure equalization sets (syringe filter 0.2 µm) is possible. It is safe to transfer of liquid media within a closed and sterile system (evaporation is reduced).
Certifications: USP standard.
Related Information
Certificates
Search by Lot NumberDWK DURAN® Innovative Bottle Systems
Product Literature
DWK Life Sciences Return to Research Promo 2021
Promotion Material
Customers Who Bought This Also Bought