ContactUsLinkComponent
Extraction HEPA/Carbon Fume Hoods, DFMZ
Extraction HEPA/Carbon Fume Hoods, DFMZ
Supplier:  Microzone
Certificates
Extraction HEPA/Carbon Fume Hoods, DFMZ
Supplier:  Microzone
Certificates
Specifications
  • Catalog No:
  • CA97015-488
  • CA97015-486
  • CA97015-484
  • Dimensions:
  • 68W×44H×23"D
  • 44W×44H×23"D
  • 32W×44H×23"D
  • Filter dimensions:
  • HEPA Filter: 23⁵/₈×11⁵/₈×3¹/₁₆"
    Carbon Filter: 23⁵/₈×11⁵/₈×1⁷/₈"
  • HEPA Filter: 35⁵/₈×11⁵/₈×3¹/₁₆"
    Carbon Filter: 17³/₄×11⁵/₈×1⁷/₈"
  • HEPA Filter: 23⁵/₈×11⁵/₈×3¹/₁₆"
    Carbon Filter: 17³/₄×11⁵/₈×1⁷/₈"