About this item
LabStrong Fi-Streem™ stills consistently produce high quality ASTM type II water and can remove inorganic ions, dissolved gases, organics, particles, bacteria, and pyrogens from water.
- Included bracket for wall or bench mounting
- Easy cleaning
- Dual feed capabilities for tap or pretreated feedwater
- Feed water operational temperature range: 40 to 120° F
- Feed water cooling water temperature: 40 to 86° F
- Cooling water: 30 LPH
The still features a unique patented vapor trap designed to eliminate all vapor borne droplets, ensuring there is no carry-over of impurities in the distillate water. The resultant is pyrogen free water at all times. Glass components in it eliminate leaching impurities.
An over temperature protection device in the boiler assembly offers protection against low water conditions. The totally enclosed still design assures operator safety from breakage and hot surfaces.
It is supplied with a float switch which shuts off the still automatically when the water storage reservoir is full, enabling safe, user friendly and unattended operation. The 24-hour automatic control activates the still as required to maintain a full reservoir.