Fractional Distillation Apparatus
Catalog #: 470310-688
Supplier: Wards
Specifications
- Description:Fractional Distillation Apparatus
- Cat. no.:470310-688
Specifications
About this item
Fractional distillation process is the separation of a mixture into individual parts of fractions by heating it up.
- All pieces are made of high quality borosilicate glass 3.3
Delivery: Kit includes 250 ml Erlenmeyer flask (1), 250 ml round bottom boiling flask (1), Vigreux column (1), Liebig condenser (1), receiver adapter vacuum (1), joint clips (4). Clamps, stand, thermometer or Bunsen burner are not included with this kit.
Customers Who Bought This Also Bought