ContactUsLinkComponent
Simple Distillation Apparatus
Simple Distillation Apparatus
Catalog #: 470310-686
Supplier:  Wards
Apparatus Simple Distillation with joint size 19/26
Simple Distillation Apparatus
Catalog #: 470310-686
Supplier:  Wards
Specifications
  • Description:
    Simple Distillation Apparatus
  • Cat. no.:
    470310-686
Specifications
About this item
This simple distillation apparatus is made up of boroscilicate glass.
Delivery: Apparatus includes round bottom flask (500 ml), still head with thermometer support, Liebig condensor (300 mm), receiver adapter and 3×joint clip (19/26).
Customers Who Bought This Also Bought