High Range Chlorine ColorQ, LaMotte®
Catalog #: 470308-894
Supplier: LaMotte
CAS Number:
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:Chlorine Test Tablets, DPD#1
- Size:100 tablets per box
- Cat. no.:470308-894
- No. of Tests:—
Specifications
About this item
The LaMotte ColorQ meter is a simple way to test low and high concentrations of chlorine.
- Free and Total Chlorine, 0-10 ppm
- Total Chlorine 0-700 ppm
- DPD reagents to measure low to high level tests
- No color matching
- Easy push-button control
- Water-resistant housing
- 5 minute auto-off function protects the battery life; 2AA batteries included
Delivery: ColorQ meter comes in a sturdy carrying case with all of the reagents and apparatus to do 50 tests each.
Customers Who Bought This Also Bought