KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Screw Cap, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Amber, screw thread vials with attached black phenolic caps and cone-shaped polyethylene liners.
Manufactured from 51 expansion borosilicate amber glass conforming to USP Type I requirements.
Packaging: Packaged in corrugated trays with partitions.
Customers Who Bought This Also Bought