Order Entry
ContactUsLinkComponent
Microplate Sealing Tape, MP Biomedicals
Microplate Sealing Tape, MP Biomedicals
Catalog #: IC097742000
Supplier:  MP Biomedicals
Acetate Sealing Tape (60m)
Microplate Sealing Tape, MP Biomedicals
Catalog #: IC097742000
Supplier:  MP Biomedicals

Specifications

  • L×W:
    60 m × 8 cm
  • Material:
    Acetate
  • Cat. no.:
    IC097742000

Specifications

About this item

Acetate plate sealers are not gas permeable and not autoclavable.

  • This microplate sealing tape is easy to apply and remove from microplates
  • Microplate sealing tape is suitable for all microplates for both short and long term storage
  • Provides a tight seal to minimize evaporation and condensation
  • 60 m per roll
Caution: Store at room temperature.
Customers Who Bought This Also Bought