Ward's Artic Squares® Freezer Rack
Catalog #: 470311-190
Supplier: VWR International
Specifications
- Capacity:16 Boxes
- Dimensions:13.9×57.1×23.8 cm
- No. of Shelves:4
Specifications
About this item
Innovative horizonal drawer system for upright freezers.
- Constructed of corrosion-resistant stainless steel with four drawers to hold 16 boxes
- Fits most standard upright freezers
The ward's artic Squares® freezer rack features drawers that have a unique stop, so that when fully extended they pivot down allowing for easy box access. The polycarbonate drawer handles provide a user-friendly grip.
Each rack also includes a slot and label for drawer/sample identification. The freezer track also includes a pull-out wire handle for easy removal off the freezer shelf.
Customers Who Bought This Also Bought