Carson 20x Microscope with Universal Smartphone Clip
Catalog #: 470311-278
Supplier: CARSON OPTICAL
Specifications
- Description:MicroMini™ Microscope with Smartphone Adapter Clip
- Cat. no.:470311-278
Specifications
About this item
Will fit most smartphones, including the iPhone XS Max/X and the latest Samsung phones.
- Features three different lights
- LED microscope light
- LED flashlight
- UV light
The MicroMini™ (MM-380) is a 20× microscope with a smartphone adapter clip. The microscope magnetically attaches to the adapter clip and is automatically aligned to the camera hole of the clip. Simply mount the adapter clip onto your smartphone camera. You will now be able to take photos and videos through your smartphone of anything you view under the microscope.
Customers Who Bought This Also Bought