To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Species:Mouse
- Size:5 mg
- Storage conditions:Store at 2-8 °C.
- Protein/peptide name:IgG1 (kappa) MOPC 21
- Formulation:Ascites
- Cat. no.:IC0850341
Specifications
About this item
[MOPC 21]
Ascites fluid, collected from Balb/c mice which carry plasmacytomas subcutaneously, are clarified by centrifugation and filtration. It is dialyzed, adjusted to yield at least 5 mg of immunoglobulin, then vialed and lyophilized. Clarified ascites are tested for immunoglobulin by immunoelectrophoresis using antisera specific for the particular type and isotype. Approximately 10% of the total protein is immunoglobulin (including trace levels of the host's immunoglobulins), 80% is albumin, and 10% is complement and other proteins.