Streptomyces hyalurolyticus Hyaluronidase
Catalog #: IC08320421
Supplier: MP Biomedicals
CAS Number:
Specifications
- Species:Streptomyces hyalurolyticus
- Size:4×100 U
- Storage conditions:Store at 0...5 °C
- Enzyme name:Hyaluronidase
- Cat. no.:IC08320421
Specifications
About this item
- Lyophilized, salt-free
- Activity: ∼2000 units/mg
- Protease and lysozyme free
- Each vial contains 100 U
Enzyme activity is determined by a change in optical density in a hyaluronic acid solution. It is compared to a hyaluronidase International Standard expressed as I.U.
Customers Who Bought This Also Bought